Order Entry
Czech Republic
ContactUsLinkComponent
Chromium(II) chloride for synthesis, Sigma-Aldrich®
Irritant
Chromium(II) chloride for synthesis, Sigma-Aldrich®
Catalog # 8.02482.0010
Supplier:  Merck
CAS Number:  
Irritant
Chromium(II) chloride for synthesis, Sigma-Aldrich®
Catalog # 8.02482.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    823 - 825 °C
  • Molar Mass:
    122.9 g/mol
  • Boiling Point:
    1300 °C

Specification Test Results

About this item

 :
 :122.9 g/mol
Merck Index:13,02264
Boiling Pt:1302 °C (1013 hPa)
Melting Pt:823…825 °C
Density:2.87 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-163-3
UN:3260
ADR:8,III
 :M-Clarity™ quality level = MQ200
 :GHS07
CAS Number:10049-05-5
Formula:Cl₂Cr
MDL Number:MFCD00010947