Tantalum(V) oxide ≥99.9% (metals basis), sputtering target 76.2 mm (3.0 in) Ø x 3.18 mm (0.125 in) thick
Catalog # 41103.KS
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
Merck Index:14,09056
Melting Pt:1870…1880 °C
Density:8.2 g/cm³ (20 °C)
:441.89 g/mol
EINECS:215-238-2
CAS Number:1314-61-0
Formula:O₅Ta₂
MDL Number:MFCD00011254