Titanium(IV) oxide rutile, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1317-80-2
:
Formula:O₂Ti
MDL Number:MFCD00011269
:79.88 g/mol
Melting Pt:1855 °C
Density:4.197 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ100
EINECS:215-282-2
Frequently Bought Together