Catalase (from Corynebacterium glutamicum)
Supplier: Merck
Certificates
About this item
- Unit definition: One unit corresponds to the amount of enzyme which decomposes 1 μmol H2O2 per minute at pH 7,0 and 25 °C
- Contains: ~30% glycerol, 10% ethanol
Specifications
- CAS No.:9001-05-2
- Source:Corynebacterium glutamicum
- Storage conditions:2...8 °C
- Form:Deep brown Solution
- Enzyme name:Catalase
- Concentration:≥500000 U/ml
Specifications
Frequently Bought Together