
Titanium(IV) oxide anatase catalyst support
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1317-70-0
:13463-67-7
Formula:TiO₂
MDL Number:MFCD00011269
:79.88 g/mol
Merck Index:13,09549
Boiling Pt:2900 °C (1013 hPa)
Melting Pt:1855 °C
Density:4.2 g/cm³ (20 °C)
EINECS:215-280-1
:GHS08
Storage Temperature:Ambient
Related Information
Frequently Bought Together