Order Entry
Czech Republic
ContactUsLinkComponent
Titanium(IV) oxide 98.0-100.5%
Titanium(IV) oxide 98.0-100.5%
Certificates
undefined
Titanium(IV) oxide 98.0-100.5%

Specification Test Results

  • Appearance:
    White to almost white Powder
  • Acidity/alkalinity:
    Passes test
  • Titration:
    98.0 to 100.5 %
  • Antimony (Sb):
    ≤100 ppm
  • Arsenic (As):
    ≤5 ppm
  • Barium (Ba):
    passes test
  • Iron (Fe):
    ≤200 ppm
  • Soluble matter:
    ≤0.5 % (in water)

Specification Test Results

Related Information

Frequently Bought Together