Order Entry
Czech Republic
ContactUsLinkComponent
Titanium nitride ≥99.5% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Titanium nitride ≥99.5% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41110.KS
CAS Number:  
Titanium nitride ≥99.5% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41110.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:25583-20-4
 :
Formula:NTi
MDL Number:MFCD00049596
 :61.89 g/mol
Melting Pt:2950 °C
Density:5.22
Storage Temperature:Ambient
EINECS:247-117-5