Titanium nitride ≥99.7% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25583-20-4
:
Formula:NTi
MDL Number:MFCD00049596
:61.89 g/mol
Melting Pt:2950 °C
Density:5.22
Storage Temperature:Ambient
EINECS:247-117-5
Related Information
Frequently Bought Together