
Hafnium(IV) chloride 99% -80 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13499-05-3
:
Formula:Cl₄Hf
MDL Number:MFCD00011034
:320.3 g/mol
Melting Pt:319 °C (subl.)
Storage Temperature:Ambient
EINECS:236-826-5
UN:3260
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White Powder
- Titration Argentometric:≥98.5 %
Specification Test Results
Frequently Bought Together