Diyttrium trioxide, Sigma-Aldrich®
Catalog # SIAL544892-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Boiling Pt:4300 °C (1013 hPa)
Melting Pt:2410 °C
Density:5.01 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:225.81 g/mol
EINECS:215-233-5
CAS Number:1314-36-9
Formula:Y₂O₃
MDL Number:MFCD00011473