Diyttrium trioxide, Supelco®
Catalog # 1.12412.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:2410 °C
- Molar Mass:225.81 g/mol
- Density:5.0 g/cm3 (20 °C)
Specification Test Results
About this item
:
Boiling Pt:4300 °C (1013 hPa)
Melting Pt:2410 °C
Density:5.01 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
:225.81 g/mol
EINECS:215-233-5
CAS Number:1314-36-9
Formula:Y₂O₃
MDL Number:MFCD00011473