
Diyttrium trioxide ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1314-36-9
:
Formula:Y₂O₃
MDL Number:MFCD00011473
Merck Index:14,10107
Boiling Pt:4300 °C (1013 hPa)
Melting Pt:2410 °C
Density:5.01
:225.81 g/mol
EINECS:215-233-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together