
Diyttrium trioxide ≥99.9% (Y basis)
Catalog # ABCRAB111934.0500
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
Specifications
About this item
:
Boiling Pt:4300 °C (1013 hPa)
Melting Pt:2410 °C
Density:5.01
:225.81 g/mol
EINECS:215-233-5
:GHS07
CAS Number:1314-36-9
Formula:Y₂O₃
MDL Number:MFCD00011473