ContactUsLinkComponent
Discandium trioxide ≥99.998% (metals basis), REacton® (REM ≤ 50 ppm)
  10916.06
 :  
Discandium trioxide ≥99.998% (metals basis), REacton® (REM ≤ 50 ppm)
  10916.06
 :  
  • Pack type:
    Plastic bottle for solids
  • Pk:
    5 g
 :
 :137.91 g/mol
 :14,8392
 :2485 °C
 :3.86 g/cm³ (20 °C)
 :Ambient
 :235-042-0
 :12060-08-1
 :O₃Sc₂
 :MFCD00011223