Zinc(II) telluride, Sigma-Aldrich®
Catalog # SIAL254320-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:192.99 g/mol
Merck Index:12,10297
Melting Pt:1294.85 °C
Density:∼6.3 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:215-260-2
CAS Number:1315-11-3
Formula:TeZn
MDL Number:MFCD00043079