Order Entry
Czech Republic
ContactUsLinkComponent
Mercury(II) sulphate ACS analytical reagent, Supelco®
Toxic
Health
Environment
Mercury(II) sulphate ACS analytical reagent, Supelco®
Supplier:  Merck
Certificates
Toxic
Health
Environment
Mercury(II) sulphate ACS analytical reagent, Supelco®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Molar Mass:
    296.65 g/mol
  • Bulk Density:
    670 kg/m3
  • Density:
    6.47 g/cm3 (20 °C)
  • pH:
    1 (50 g/l, H2O, 20 °C)
  • Ignition Point:
    450 °C
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic materials

Specification Test Results

Related Information

Frequently Bought Together