Order Entry
Czech Republic
ContactUsLinkComponent
Mercury(II) sulphate 99+%
Toxic
Health
Environment
Mercury(II) sulphate 99+%
Certificates
undefined
Toxic
Health
Environment
Mercury(II) sulphate 99+%

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Assay:
    ≥99.0 %

Specification Test Results

Related Information

Frequently Bought Together