Order Entry
Czech Republic
ContactUsLinkComponent
Mercury (II) oxide, red ≥99.0% ACS, Fluka™
Health
Toxic
Environment
Mercury (II) oxide, red ≥99.0% ACS, Fluka™
Catalog # 213357-100G
CAS Number:  
Health
Toxic
Environment
Mercury (II) oxide, red ≥99.0% ACS, Fluka™
Catalog # 213357-100G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Insoluble in HCl:
    Max. 0.03 %
  • Residue on ignition a. reducing with HCOOH (SO₄)*:
    Max. 0.025 %
  • Iron (Fe):
    Max. 0.005 %
  • Chloride (Cl):
    Max. 0.025 %
  • Sulfate (SO₄):
    Max. 0.015 %
  • Total N:
    Max. 0.005 %

Specification Test Results

About this item

 :
 :216.59 g/mol
Merck Index:13,05908
Boiling Pt:356,6 °C (1013 hPa)
Melting Pt:400 °C (dec.)
Density:11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:244-654-7
UN:1641
ADR:6.1,II
 :GHS08
 :GHS06
 :GHS09
CAS Number:21908-53-2
Formula:HgO
MDL Number:MFCD00011045