Order Entry
Czech Republic
ContactUsLinkComponent
Mercury (II) oxide, yellow ≥99.0% ACS, Fluka™
Health
Toxic
Environment
Mercury (II) oxide, yellow ≥99.0% ACS, Fluka™
Certificates
Health
Toxic
Environment
Mercury (II) oxide, yellow ≥99.0% ACS, Fluka™
Certificates

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Glowing After reducing with HCOOH(as SO₄):
    Max. 0.05 %
  • Insoluble in HCl:
    Max. 0.03 %
  • Iron (Fe):
    Max. 0.003 %
  • Chloride (Cl):
    Max. 0.025 %
  • Sulfate (SO₄):
    Max. 0.01 %
  • Total N:
    Max. 0.005 %

Specification Test Results

Related Information

Frequently Bought Together