


Mercury(II) iodide, red Ph. Franç., extra pure, Supelco®
Catalog # 1.04420.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Solubility:0.06 g/l (25 °C)
- Melting Point:259 °C
- Molar Mass:454.39 g/mol
- Bulk Density:1350 kg/m3
- Boiling Point:354 °C (1013 hPa) (sublimed)
- Vapor Pressure:0.006 hPa (80 °C)
- Density:6.36 g/cm3 (25 °C)
- pH:6 - 7 (50 g/l, H2O, 20 °C) (slurry)
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Specification Test Results
About this item
:
:454.4 g/mol
Merck Index:13,05905
Boiling Pt:354 °C (1013 hPa)
Melting Pt:259 °C
Density:6.36 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-873-8
UN:1638
ADR:6.1,II
:M-Clarity™ quality level = MQ200
:GHS06
:GHS08
:GHS09
CAS Number:7774-29-0
Formula:HgI₂
MDL Number:MFCD00011044