


Cadmium(II) iodide, Sigma-Aldrich®
Catalog # SIAL20910-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:366.22 g/mol
Merck Index:12,01657
Boiling Pt:787 °C (1013 hPa)
Melting Pt:387 °C
Density:5.67 g/cm³ (20 °C)
EINECS:232-223-6
UN:2570
ADR:6.1,III
:GHS06
:GHS08
:GHS09
CAS Number:7790-80-9
Formula:CdI₂
MDL Number:MFCD00010920