


Cadmium(II) iodide ≥99.5% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7790-80-9
:
Formula:CdI₂
MDL Number:MFCD00010920
:366.22 g/mol
Merck Index:14,1621
Boiling Pt:787 °C
Melting Pt:387…389 °C
Density:5.67
Storage Temperature:Ambient
EINECS:232-223-6
UN:2570
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Related Information
Frequently Bought Together