


Zinc chloride, anhydrous ≥99.95% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7646-85-7
:
Formula:ZnCl₂
MDL Number:MFCD00011295
:136.3 g/mol
Merck Index:14,10132
Boiling Pt:732 °C
Melting Pt:290 °C
Density:2.91
Storage Temperature:Ambient
EINECS:231-592-0
UN:2331
ADR:8,III
:GHS05
:GHS07
:GHS09
Related Information
Frequently Bought Together