Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
 
 
Mercury(II) chloride 99.5+% ACS
 
undefined
 
 
 
 
Mercury(II) chloride 99.5+% ACS
 

 

  • Appearance:
    White Crystalline powder or crystals
  • Titration Complexometric:
    ≥99.5 %
  • Solubility:
    (in ethyl ether) Passes test
  • Iron (Fe):
    ≤20 ppm
  • Residue after reduction:
    ≤0.02 %

 

 

Frequently Bought Together