Cadmium(II) chloride, Sigma-Aldrich®
: Merck
:10108-64-2
:
:CdCl₂
:MFCD00010916
:183.32 g/mol
:12,01653
:967 °C (1013 hPa)
:565 °C
:4.05 g/cm³ (20 °C)
:2570
:6.1,III
:M-Clarity™ quality level = MQ100
:233-296-7
:GHS06
:GHS08
:GHS09
Frequently Bought Together