Cadmium(II) chloride, ultra dry ≥99.996% (metals basis)
:10108-64-2
:
:CdCl₂
:MFCD00010916
:183.32 g/mol
:14,1617
:960 °C
:568 °C
:4.047
:Ambient
:2570
:6.1,III
:ampouled under argon
:233-296-7
:GHS06
:GHS08
:GHS09
Frequently Bought Together