


Cadmium(II) chloride, ultra dry ≥99.998% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10108-64-2
:
Formula:CdCl₂
MDL Number:MFCD00010916
:183.32 g/mol
Merck Index:14,1617
Boiling Pt:960 °C
Melting Pt:568 °C
Density:4.047
Storage Temperature:Ambient
UN:2570
ADR:6.1,III
:ampouled under argon
EINECS:233-296-7
:GHS06
:GHS08
:GHS09
Related Information
Frequently Bought Together