Dehydrated culture media, ITC broth base
Katalogové číslo SIAL17156-500G
Dodavatel: Merck
Číslo CAS:
Specifikace
- Pk:500 g
- Description:ITC broth base
Specifikace
O tomto produktu
ITC broth base can be used for the selective enrichment, enumeration of Y. enterocolitica.
- Final pH: 6,9 ±0,2 (25 °C)
Certifications: Meets the specification of ISO 10273:2003.