
Copper(I) iodide ≥99.998% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7681-65-4
:1335-23-5
Formula:CuI
MDL Number:MFCD00010978
:190.45 g/mol
Merck Index:14,2662
Boiling Pt:∼1290 °C
Melting Pt:605 °C
Density:5.63
Storage Temperature:Ambient
EINECS:231-674-6
UN:3077
ADR:9,III
:GHS07
Frequently Bought Together