Order Entry
Czech Republic
ContactUsLinkComponent
Copper(I) chloride, EMSURE® ACS for analysis, Supelco®
Copper(I) chloride, EMSURE® ACS for analysis, Supelco®
Catalog # 1.02739.0250
Supplier:  Merck
CAS Number:  
Copper(I) chloride, EMSURE® ACS for analysis, Supelco®
Catalog # 1.02739.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Solubility:
    0.001 - 0.1 g/l
  • Melting Point:
    422 °C
  • Molar Mass:
    99 g/mol
  • Bulk Density:
    1600 - 1800 kg/m3
  • Boiling Point:
    1367 °C
  • Density:
    4.14 g/cm3 (25 °C)
  • pH:
    5 (50 g/l, H2O, 20 °C) (slurry)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

About this item

 :
Merck Index:13,02690
Boiling Pt:1367 °C (1013 hPa)
Melting Pt:430 °C
Density:4.145 g/cm³ (25 °C)
ADR:8,III
 :M-Clarity™ quality level = MQ300
 :99 g/mol
EINECS:231-842-9
UN:2802
 :GHS05
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:7758-89-6
Formula:ClCu
MDL Number:MFCD00010971