ContactUsLinkComponent
Copper(II) bromide, anhydrous 99+%, extra pure
Irritant
Corrosive
Environment
Copper(II) bromide, anhydrous 99+%, extra pure
Certificates
undefined
Irritant
Corrosive
Environment
Copper(II) bromide, anhydrous 99+%, extra pure
Specification Test Results
  • Appearance:
    Dark grey to grey-black Crystalline powder
  • Titration Iodometric:
    ≥99.0 %
  • Chloride (Cl):
    ≤5000 ppm
  • Sulfate (SO4):
    ≤2000 ppm
  • Insoluble matter:
    ≤0.05 % (in water)