(3-Acrylamidopropyl)trimethylammonium chloride ≥75% (w/w) in water stabilised, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:45021-77-0
:
Formula:C₉H₁₉ClN₂O
MDL Number:MFCD00274249
:206.71 g/mol
Density:1.11 g/ml (25 °C)
:M-Clarity™ quality level = MQ200
:Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) 3000 ppm
Storage Temperature:Refrigerator
EINECS:256-181-3
Specification Test Results
- Infrared spectrum:Conforms to structure
- Titration by AgNO₃:73.0 - 77.0%
- Concentration:75 wt. % in H₂O
Specification Test Results
Frequently Bought Together