1,1'-Binaphthyl 97%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1,1'-Binaphthalene
- CAS Number:604-53-5
- Formula:C₂₀H₁₄
- MDL Number:MFCD00041740
- Molecular Weight:254.33 g/mol
- Boiling Pt:405 °C (1013 hPa)
- Melting Pt:143…146 °C
- Density:1,19 g/cm³ (20 °C)
- EINECS:210-070-6
Documents
ACRO40224
SDS
Specifications
Specification Test Results
- Appearance:White to brown Powder
- Infrared spectrum:Conforms
- GC:≥96.0 %
- Titration with HClO4:≥97.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



