
1,7-Dihydroxynaphthalene, Sigma-Aldrich®
Catalog # SIAL535486-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:160.17 g/mol
Boiling Pt:375 °C (1013 hPa)
Melting Pt:175…177 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:209-383-0
:GHS07
CAS Number:575-38-2
Formula:C₁₀H₈O₂
MDL Number:MFCD00035720