Order Entry
Czech Republic
ContactUsLinkComponent
1,4-Dihydroxynaphthalene, Sigma-Aldrich®
1,4-Dihydroxynaphthalene, Sigma-Aldrich®
Catalog # SIAL70430-10G
Supplier:  Merck
CAS Number:  
1,4-Dihydroxynaphthalene, Sigma-Aldrich®
Catalog # SIAL70430-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :160.17 g/mol
Melting Pt:191…193 °C
Density:1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-336-4
CAS Number:571-60-8
Formula:C₁₀H₈O₂
MDL Number:MFCD00003977