Order Entry
Czech Republic
ContactUsLinkComponent
1,3-Dihydroxynaphthalene, Supelco®
Irritant
1,3-Dihydroxynaphthalene, Supelco®
Catalog # 70650-5G
Supplier:  Merck
CAS Number:  
undefined
Irritant
1,3-Dihydroxynaphthalene, Supelco®
Catalog # 70650-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :160.17 g/mol
Merck Index:12,06483
Boiling Pt:386 °C (1013 hPa)
Melting Pt:124 °C
Density:1.12 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:205-079-7
 :GHS07
CAS Number:132-86-5
Formula:C₁₀H₈O₂
MDL Number:MFCD00003965