Order Entry
Czech Republic
ContactUsLinkComponent
2,7-Dimethoxynaphthalene, Sigma-Aldrich®
2,7-Dimethoxynaphthalene, Sigma-Aldrich®
Catalog # SIAL213152-5G
Supplier:  Merck
CAS Number:  
2,7-Dimethoxynaphthalene, Sigma-Aldrich®
Catalog # SIAL213152-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :188.23 g/mol
Boiling Pt:299 °C (1013 hPa)
Melting Pt:144 °C
 :M-Clarity™ quality level = MQ100
EINECS:222-433-6
CAS Number:3469-26-9
Formula:C₁₂H₁₂O₂
MDL Number:MFCD00004064