
1,3-Dinitronaphthalene, Sigma-Aldrich®
Catalog # SIAL128066-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:218.17 g/mol
Boiling Pt:377 °C (1013 hPa)
Melting Pt:148 °C
:M-Clarity™ quality level = MQ100
EINECS:210-116-5
:GHS07
CAS Number:606-37-1
Formula:C₁₀H₆N₂O₄