
2-Naphthoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:93-09-4
:
Formula:C₁₁H₈O₂
MDL Number:MFCD00004101
:172.18 g/mol
Merck Index:14,6382
Boiling Pt:333 °C (1013 hPa)
Melting Pt:182…186 °C
Density:1.08
Flash Pt:205 °C (401 °F)
EINECS:202-217-8
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together