Order Entry
Czech Republic
ContactUsLinkComponent
Ammonium cerium(IV) sulphate dihydrate ≥98.0% (RT), Puriss. p.a. ACS analytical reagent, Fluka™
Ammonium cerium(IV) sulphate dihydrate ≥98.0% (RT), Puriss. p.a. ACS analytical reagent, Fluka™
Catalog # 22269-500G
CAS Number:  
Ammonium cerium(IV) sulphate dihydrate ≥98.0% (RT), Puriss. p.a. ACS analytical reagent, Fluka™
Catalog # 22269-500G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay (Iodometric):
    98.0 - 102.0 %
  • Insoluble in acid:
    Max. 0.05 %
  • Iron (Fe):
    Max. 100 ppm
  • Phosphate (PO₄):
    Max. 300 ppm

Specification Test Results

About this item

 :
 :632.55 g/mol
Melting Pt:130 °C
Storage Temperature:Ambient
EINECS:231-567-4
CAS Number:10378-47-9
Formula:(NH₄)₂Ce(SO₄)₄·2H₂O
MDL Number:MFCD00149417