Pentaerythritol tetrakis [3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] ≥98% (by TLC), Sigma-Aldrich®
Catalog # SIAL441783-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Poly bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Purity (TLC):≥98%
- Infrared spectrum:Conforms to structure
Specification Test Results
About this item
:
:1177.66 g/mol
Melting Pt:115…118 °C (dec., lit.)
:M-Clarity™ quality level = MQ200
EINECS:229-722-6
CAS Number:6683-19-8
Formula:C₇₃H₁₀₈O₁₂
MDL Number:MFCD00059345