Order Entry
Czech Republic
ContactUsLinkComponent
Ammonium cerium(IV) sulphate dihydrate, EMSURE® for analysis, Supelco®
Ammonium cerium(IV) sulphate dihydrate, EMSURE® for analysis, Supelco®
Catalog # 1.02273.0100
Supplier:  Merck
CAS Number:  
Ammonium cerium(IV) sulphate dihydrate, EMSURE® for analysis, Supelco®
Catalog # 1.02273.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    (20 °C) Hydrolysis
  • Molar Mass:
    632.55 g/mol
  • Bulk Density:
    800 kg/m3
  • pH:
    1.2 (100 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :632.55 g/mol
Melting Pt:130 °C
 :M-Clarity™ quality level = MQ300
EINECS:231-567-4
CAS Number:10378-47-9
Formula:(NH₄)₂Ce(SO₄)₄·2H₂O
MDL Number:MFCD00149417