
Lutetium(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12032-20-1
:
Formula:Lu₂O₃
MDL Number:MFCD00011100
:397.93 g/mol
Merck Index:14,5615
Melting Pt:2487 °C
Density:9.420
EINECS:234-764-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together