
Neodymium trichloride hexahydrate, Sigma-Aldrich®
Catalog # SIAL289183-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:358.69 g/mol
Melting Pt:123 °C
Density:2.28 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:233-031-5
:GHS07
CAS Number:13477-89-9
Formula:Cl₃Nd·6H₂O
MDL Number:MFCD00149801