Order Entry
Czech Republic
ContactUsLinkComponent
2-Phenylimidazole 98%
Irritant
2-Phenylimidazole 98%
Certificates
undefined
Irritant
2-Phenylimidazole 98%

Specification Test Results

  • Appearance:
    Light yellow to beige Crystalline or granular powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    142 to 148 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together