1,1'-Carbonyldiimidazole, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Di(1H-imidazol-1-yl)methanone, CDI Crosslinker, 1,1'-Carbonylbis(imidazole), 1,1'-Carbonylbis-1H-imidazole, N,N'-Carbonyldiimidazole, CDI
CAS Number:530-62-1
:
Formula:C₇H₆N₄O
MDL Number:MFCD00005286
Molecular Weight:162.15 g/mol
Melting Pt:111…116 °C
ADR:8 II
Merck Index:14,01819
:M-Clarity™ quality level = MQ300
EINECS:208-488-9
UN:3263
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together