4-Methylimidazole, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:822-36-6
:
Formula:C₄H₆N₂
MDL Number:MFCD00005201
:82.11 g/mol
Boiling Pt:263 °C (1013 hPa)
Density:1.0416 g/cm³ (14.3 °C)
EINECS:212-497-3
UN:3263
ADR:8,III
:M-Clarity™ quality level = MQ100
Frequently Bought Together