Order Entry
Czech Republic
ContactUsLinkComponent
4-Methylimidazole 98%
Toxic
4-Methylimidazole 98%
Certificates
undefined
Toxic
4-Methylimidazole 98%

Specification Test Results

  • Appearance:
    White to yellow Crystalline powder or crystals or mass and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    50 to 56 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together