Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Imidazole

Imidazole

Plastic bottle for solids, 250 g
Catalog # A3635.0250
Corrosive
Health
Irritant
Supplier:   PanReac AppliChem
Imidazole
Catalog # A3635.0250
Supplier:  PanReac AppliChem
CAS Number:  288-32-4
Imidazole
Imidazole

Plastic bottle for solids, 250 g

CAS Number:  288-32-4
Price
45 010,00 Kč
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Glyoxaline, 1H-Imidazole, Glyoxalamine
  • CAS Number:288-32-4
  • Formula:C₃H₄N₂
  • MDL Number:MFCD00005183
  • Molecular Weight:68.08 g/mol
  • Merck Index:13,04935
  • Boiling Pt:255 °C (1013 hPa)
  • Melting Pt:89…90 °C
  • Density:1.18 g/cm³ (20 °C)
  • Flash Pt:135 °C
  • Storage Temperature:Ambient
  • EINECS:206-019-2
  • UN:2923
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g

Specifications