Order Entry
Czech Republic
ContactUsLinkComponent
Phenylhydrazine ≥97.0% for synthesis, Sigma-Aldrich®
Toxic
Health
Environment
Phenylhydrazine ≥97.0% for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
Health
Environment
Phenylhydrazine ≥97.0% for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (HClO₄):
    ≥97.0%
  • Density (d 20 °C/ 4 °C):
    1.096 - 1.099
  • Identity (IR):
    Passes Test
  • Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.:
    244 °C (1013 hPa)
  • Vapor Pressure:
    0.1 hPa (20 °C)
  • Flash Point:
    88 °C
  • Refractive Index:
    1.607 (20 °C, 589 nm)
  • Explosion Limit:
    1.1 %(V)
  • Density:
    1.10 g/cm3 (20 °C)
  • pH:
    (H2O, 25 °C) alkaline,(undiluted)
  • Ignition Point:
    195 °C

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together